siRNA Details
Gene Symbol: Tgfb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009368.3 | NP_033394.2 | 2 | 1493 | ||
GenBank mRNA | AK132519.1 | - | 2 | 1483 | ||
AK145304.1 | - | 2 | 1478 | |||
AK148385.1 | - | 2 | 1357 | |||
AK162560.1 | - | 2 | 257 | |||
BC005513.1 | - | 2 | 235 | |||
BC014690.1 | - | 2 | 772 | 3663150 | ||
BC089347.1 | - | 2 | 1450 | |||
BC094591.1 | - | 3 | 918 | |||
BC108426.1 | - | 2 | 1493 | 6757707 | ||
M32745.1 | - | 2 | 988 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
