siRNA Details
Gene Symbol: Tgfb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009368.3 | NP_033394.2 | 4 | 1824 | ||
GenBank mRNA | AK132519.1 | - | 4 | 1814 | ||
AK145304.1 | - | 4 | 1809 | |||
AK148385.1 | - | 4 | 1688 | |||
AK161328.1 | - | 1 | 71 | |||
AK162560.1 | - | 4 | 588 | |||
BC005513.1 | - | 4 | 566 | |||
BC014690.1 | - | 4 | 1103 | 3663150 | ||
BC089347.1 | - | 4 | 1781 | |||
BC094591.1 | - | 5 | 1249 | |||
BC108426.1 | - | 4 | 1824 | 6757707 | ||
M32745.1 | - | 4 | 1319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
