siRNA Details
Gene Symbol: Klf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289471.1 | NP_001276400.1 | 3 | 968 | ||
NM_013692.3 | NP_038720.1 | 3 | 968 | |||
GenBank mRNA | AF049879.1 | - | 3 | 964 | ||
AF064088.1 | - | 3 | 960 | |||
AK004906.1 | - | 3 | 967 | |||
AK043433.1 | - | 3 | 964 | |||
AK079679.1 | - | 3 | 966 | |||
AK145676.1 | - | 3 | 841 | |||
AK154719.1 | - | 3 | 964 | |||
BC003316.1 | - | 3 | 908 | 3155247 | ||
Z36270.1 | - | 3 | 968 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
