siRNA Details
Gene Symbol: Ska3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198605.3 | NP_941007.1 | 3 | 336 | ||
GenBank mRNA | AK089223.1 | - | 3 | 337 | ||
AK158142.1 | - | 3 | 358 | |||
AK166171.1 | - | 3 | 327 | |||
BC117500.1 | - | 3 | 328 | 40105921 | ||
BC117501.1 | - | 3 | 328 | 40105920 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
