siRNA Details
Gene Symbol: Top2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011623.2 | NP_035753.2 | 16 | 2016 | ||
GenBank mRNA | AK033321.1 | - | 4 | 398 | ||
D12513.1 | - | 16 | 1942 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
