siRNA Details
Gene Symbol: Xntrpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011644.3 | NP_035774.2 | 14, 15 | 2516 | ||
GenBank mRNA | AK040538.1 | - | 14, 15 | 2857 |
Gene Symbol: Trpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109897.2 | NP_001103367.1 | 5, 6 | 1352 | ||
GenBank mRNA | AF230803.1 | - | 5, 6 | 1352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
