siRNA Details
Gene Symbol: Vdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011695.2 | NP_035825.1 | 7 | 595 | ||
GenBank mRNA | AK012359.1 | - | 7 | 593 | ||
AK078555.1 | - | 7 | 599 | |||
AK150504.1 | - | 7 | 621 | |||
AK150627.1 | - | 7 | 621 | |||
AK150940.1 | - | 7 | 613 | |||
AK152274.1 | - | 7 | 593 | |||
AK159561.1 | - | 7 | 830 | |||
AK168199.1 | - | 7 | 566 | |||
AK168328.1 | - | 7 | 592 | |||
BC003731.1 | - | 7 | 595 | 3601668 | ||
U30838.1 | - | 7 | 595 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
