siRNA Details
Gene Symbol: Ezr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009510.2 | NP_033536.2 | 3 | 289 | ||
GenBank mRNA | AK002766.1 | - | 3 | 263 | ||
AK004588.2 | - | 3 | 264 | |||
AK135194.1 | - | 3 | 265 | |||
AK135353.1 | - | 3 | 265 | |||
AK145707.1 | - | 3 | 290 | |||
AK159717.1 | - | 3 | 267 | |||
AK159824.1 | - | 3 | 265 | |||
AK159937.1 | - | 3 | 266 | |||
AK166191.1 | - | 3 | 265 | |||
AK168410.1 | - | 3 | 265 | |||
AK168840.1 | - | 3 | 265 | |||
AK170610.1 | - | 3 | 265 | |||
AK172336.1 | - | 3 | 272 | |||
BC048181.1 | - | 3 | 252 | 6826190 | ||
BC098502.1 | - | 3 | 241 | 30047137 |

More Information
Other Species Targeted

Gene Ontology Categories:
