siRNA Details
Gene Symbol: Abra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175456.4 | NP_780665.1 | 1 | 473 | ||
GenBank mRNA | AF504061.1 | - | 1 | 421 | ||
AK045186.1 | - | 302 | ||||
AK081696.1 | - | 1 | 474 | |||
AK138943.1 | - | 172 | ||||
BC119198.1 | - | 1 | 453 | 8733947 | ||
BC120842.1 | - | 1 | 453 | 40129765 |

More Information
Other Species Targeted

Gene Ontology Categories:
