siRNA Details
Gene Symbol: Zdhhc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146073.3 | NP_666185.3 | 8 | 1867 | ||
GenBank mRNA | AF542387.1 | - | 9 | 1536 | ||
AK041496.1 | - | 8 | 1550 | |||
AK046719.1 | - | 8 | 1855 | |||
AK080845.1 | - | 8 | 1871 | |||
BC059814.1 | - | 8 | 1910 | 6813525 | ||
BC086758.1 | - | 9 | 923 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
