siRNA Details
Gene Symbol: Xlr3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011727.2 | NP_035857.2 | 7 | 538 | ||
GenBank mRNA | L22977.1 | - | 523 | |||
U02600.1 | - | 7 | 538 |
Gene Symbol: Xlr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081643.1 | NP_001075112.1 | 7 | 569 | ||
GenBank mRNA | AK146398.1 | - | 6 | 1338 | ||
BC038862.1 | - | 7 | 532 | |||
BC057003.1 | - | 5 | 2732 | |||
BC064787.1 | - | 7 | 526 | 30046437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
