siRNA Details
Gene Symbol: Dlgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128180.1 | NP_001121652.1 | 9 | 1833 | ||
NM_001128181.1 | NP_001121653.1 | 10 | 1835 | |||
NM_027712.3 | NP_081988.3 | 10 | 2820 | |||
NM_177639.6 | NP_808307.2 | 11 | 2850 | |||
GenBank mRNA | AK016873.1 | - | 2 | 276 | ||
AK034182.1 | - | 5 | 709 | |||
AK035763.1 | - | 14 | 3401 | |||
AK136926.1 | - | 10 | 1796 | |||
AK147448.1 | - | 14 | 3637 | |||
AK161941.1 | - | 9 | 1808 | |||
AK220515.1 | - | 13 | 3700 | |||
AY243846.2 | - | 10 | 2601 | |||
BC062120.1 | - | 11 | 2851 | 6822047 | ||
BC094369.1 | - | 9 | 2029 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
