siRNA Details
Gene Symbol: Prdm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033281.3 | NP_001028453.1 | 5 | 1350 | ||
GenBank mRNA | AK133910.1 | - | 5 | 1351 | ||
AK135836.1 | - | 2 | 224 | |||
AK135840.1 | - | 2 | 224 | |||
BC139295.1 | - | 5 | 1380 | 8862317 | ||
DQ340564.1 | - | 5 | 926 | |||
DQ340565.1 | - | 5 | 845 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
