siRNA Details
Gene Symbol: Rnf165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164504.1 | NP_001157976.1 | 6, 7 | 849 | ||
GenBank mRNA | AK133955.1 | - | 3, 4 | 236 | ||
BB619563.1 | - | 3, 4 | 236 | |||
CF745657.1 | - | 1, 2 | 23 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
