siRNA Details
Gene Symbol: Lct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081078.1 | NP_001074547.1 | 15, 16 | 5362 | ||
GenBank mRNA | AK158042.1 | - | 9, 10 | 3464 | ||
AY751548.1 | - | 3, 4 | 463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
