siRNA Details
Gene Symbol: Zfp101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009542.2 | NP_033568.2 | 4 | 1163 | ||
GenBank mRNA | AK076355.1 | - | 4 | 1167 | ||
AK076461.1 | - | 1 | 953 | |||
AK078458.1 | - | 4 | 1151 | |||
AK169234.1 | - | 4 | 1164 | |||
U07861.1 | - | 4 | 1123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
