siRNA Details
Gene Symbol: Dars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172644.3 | NP_766232.1 | 17 | 2394 | ||
GenBank mRNA | AK030952.1 | - | 17 | 2383 | ||
AK033634.1 | - | 15 | 3405 | |||
AK037337.1 | - | 17 | 2413 | |||
AK083300.1 | - | 6 | 803 | |||
AK138809.1 | - | 17 | 2395 | |||
AK154133.1 | - | 17 | 2422 | |||
AK154348.1 | - | 17 | 2423 | |||
AK164885.1 | - | 17 | 2366 | |||
AK204411.1 | - | 3 | 326 | |||
BC046429.1 | - | 10 | 1159 | |||
BC057943.1 | - | 15 | 2543 | |||
BC076606.1 | - | 17 | 2386 | 30625515 | ||
BC089191.1 | - | 17 | 2368 | 30473852 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
