siRNA Details
Gene Symbol: Prrc2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081290.1 | NP_001074759.1 | 3 | 519 | ||
GenBank mRNA | AK007762.1 | - | 3 | 480 | ||
AK013732.1 | - | 3 | 360 | |||
AK030766.1 | - | 3 | 459 | |||
AK077748.1 | - | 3 | 360 | |||
AK082487.1 | - | 3 | 489 | |||
AK122441.1 | - | 3 | 432 | |||
AK135245.1 | - | 3 | 461 | |||
AK166509.1 | - | 3 | 508 | |||
AK167018.1 | - | 3 | 474 | |||
BC006723.1 | - | 3 | 463 | |||
BC021412.1 | - | 3 | 447 | |||
BC037745.1 | - | 3 | 444 | |||
BC053406.1 | - | 3 | 406 | |||
BC064009.1 | - | 3 | 428 | |||
BC080672.1 | - | 3 | 438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
