siRNA Details
Gene Symbol: Rcsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038846.1 | NP_001033935.1 | 5 | 1108 | ||
NM_178593.3 | NP_848708.2 | 6 | 1198 | |||
GenBank mRNA | AK133961.1 | - | 6 | 1153 | ||
AK158429.1 | - | 6 | 1139 | |||
AK168816.1 | - | 5 | 1079 | |||
AK171323.1 | - | 1088 | ||||
AK171914.1 | - | 6 | 1121 | |||
BC025872.1 | - | 6 | 1118 | 5037953 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
