siRNA Details
Gene Symbol: Zfp36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011756.4 | NP_035886.1 | 2 | 1658 | ||
GenBank mRNA | AK005006.1 | - | 1 | 2366 | ||
AK088525.1 | - | 1 | 2366 | |||
AK154825.1 | - | 2 | 1686 | |||
AK159856.1 | - | 1 | 1456 | |||
BC021391.1 | - | 2 | 1657 | 5053664 | ||
M57422.1 | - | 2 | 1657 | |||
M58565.1 | - | 2 | 1627 | |||
M58691.1 | - | 2682 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
