siRNA Details
Gene Symbol: Rpp38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013376.2 | NP_001013394.2 | 2 | 232 | ||
GenBank mRNA | AK049133.1 | - | 2 | 177 | ||
AK087782.1 | - | 2 | 233 | |||
AK139023.1 | - | 2 | 160 | |||
BC051448.1 | - | 2 | 207 | 30060460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
