siRNA Details
Gene Symbol: Vps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172269.3 | NP_758473.3 | 4 | 1866 | ||
GenBank mRNA | AK029109.1 | - | 4 | 1843 | ||
AK033333.1 | - | 4 | 1851 | |||
AK036915.1 | - | 4 | 1851 | |||
AK036947.1 | - | 1 | 29 | |||
AK039848.1 | - | 4 | 1850 | |||
AK042471.1 | - | 4 | 1854 | |||
AK044821.1 | - | 4 | 1869 | |||
AK148077.1 | - | 4 | 1858 | |||
AK151113.1 | - | 1 | 488 | |||
AK152700.1 | - | 1 | 488 | |||
AK160689.1 | - | 4 | 1844 | |||
AK173179.1 | - | 1 | 1012 | |||
BC026870.1 | - | 1 | 792 | |||
BC036129.1 | - | 4 | 1794 | 4972693 | ||
BC039176.1 | - | 4 | 1773 | 4221232 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
