siRNA Details
Gene Symbol: Vps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172269.3 | NP_758473.3 | 5 | 2553 | ||
GenBank mRNA | AK029109.1 | - | 5 | 2530 | ||
AK033333.1 | - | 5 | 2538 | |||
AK036915.1 | - | 5 | 2538 | |||
AK036947.1 | - | 2 | 716 | |||
AK039848.1 | - | 5 | 2537 | |||
AK042471.1 | - | 5 | 2541 | |||
AK148077.1 | - | 5 | 2545 | |||
AK151113.1 | - | 2 | 1175 | |||
AK152700.1 | - | 2 | 1175 | |||
AK160689.1 | - | 5 | 2531 | |||
AK173179.1 | - | 2 | 1699 | |||
AK200305.1 | - | 2 | 125 | |||
BC026870.1 | - | 2 | 1479 | |||
BC036129.1 | - | 5 | 2481 | 4972693 | ||
BC039176.1 | - | 5 | 2460 | 4221232 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
