siRNA Details
Gene Symbol: Ppp1r3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085501.2 | NP_001078970.1 | 1 | 1929 | ||
GenBank mRNA | AK011539.1 | - | 1 | 357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
