siRNA Details
Gene Symbol: Cdc14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080818.2 | NP_001074287.1 | 2 | 623 | ||
NM_001173553.1 | NP_001167024.1 | 2 | 623 | |||
GenBank mRNA | AK032515.1 | - | 1 | 1312 | ||
AK043956.1 | - | 2 | 607 | |||
AK048250.1 | - | 2 | 528 | |||
AK052560.1 | - | 2 | 614 | |||
AK136546.1 | - | 2 | 82 | |||
AK142294.1 | - | 2 | 623 | |||
BC072644.1 | - | 2 | 577 | 30356962 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
