siRNA Details
Gene Symbol: Nol6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139236.3 | NP_631982.2 | 8 | 1156 | ||
GenBank mRNA | AF361077.1 | - | 8 | 1105 | ||
AF361078.1 | - | 8 | 1105 | |||
AK089138.1 | - | 8 | 1076 | |||
AK155818.1 | - | 8 | 1095 | |||
AK166559.1 | - | 8 | 1082 | |||
AK166650.1 | - | 8 | 1082 | |||
AK171163.1 | - | 8 | 1093 | |||
AK171421.1 | - | 8 | 1083 | |||
BC059820.1 | - | 8 | 1074 | 6827596 | ||
BC060189.1 | - | 8 | 1084 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
