siRNA Details
Gene Symbol: Usp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301414.1 | NP_001288343.1 | 8 | 2080 | ||
NM_146144.4 | NP_666256.2 | 8 | 2061 | |||
GenBank mRNA | AK080882.1 | - | 8 | 2062 | ||
BC018179.1 | - | 8 | 1941 | 3585191 | ||
BC020007.1 | - | 8 | 1913 | 3980469 | ||
BC023812.1 | - | 5 | 1235 |

More Information
Other Species Targeted

Gene Ontology Categories:
