siRNA Details
Gene Symbol: Usp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301414.1 | NP_001288343.1 | 9 | 2686 | ||
NM_146144.4 | NP_666256.2 | 9 | 2667 | |||
GenBank mRNA | AK080882.1 | - | 9 | 2668 | ||
BC018179.1 | - | 9 | 2547 | 3585191 | ||
BC020007.1 | - | 9 | 2519 | 3980469 | ||
BC023812.1 | - | 6 | 1841 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
