siRNA Details
Gene Symbol: Usp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301414.1 | NP_001288343.1 | 3 | 746 | ||
NM_146144.4 | NP_666256.2 | 3 | 727 | |||
GenBank mRNA | AK080882.1 | - | 3 | 728 | ||
BC018179.1 | - | 3 | 607 | 3585191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
