siRNA Details
Gene Symbol: Zyg11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167936.1 | NP_001161408.1 | 10 | 1735 | ||
GenBank mRNA | BC022150.1 | - | 8 | 1283 |

More Information
Other Species Targeted

Gene Ontology Categories:
