siRNA Details
Gene Symbol: Pabpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130881.2 | NP_570951.2 | 15 | 2785 | ||
NM_148917.2 | NP_683717.1 | 14 | 2276 | |||
GenBank mRNA | AK016732.1 | - | 10 | 1279 | ||
AK135962.1 | - | 2254 | ||||
AK160589.1 | - | 8 | 866 | |||
AK162172.1 | - | 7 | 867 | |||
AK215417.1 | - | 2 | 178 | |||
BC010345.1 | - | 14 | 2276 | 3707766 | ||
BC056432.1 | - | 15 | 2785 | 5696369 | ||
BC058263.1 | - | 15 | 2949 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
