siRNA Details
Gene Symbol: Ncmap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168498.1 | NP_001161970.1 | 5 | 1255 | ||
NM_001168500.1 | NP_001161972.1 | 5 | 1457 | |||
NM_001243305.1 | NP_001230234.1 | 4 | 1068 | |||
NM_001243306.1 | NP_001230235.1 | 6 | 1625 | |||
NM_145555.2 | NP_663530.1 | 5 | 1454 | |||
NM_181047.1 | NP_851390.1 | 5 | 1252 | |||
GenBank mRNA | AK014757.1 | - | 5 | 1231 | ||
AK039485.1 | - | 4 | 1097 | |||
AK076488.1 | - | 5 | 1206 | |||
AK167453.1 | - | 5 | 1455 | |||
BC005730.1 | - | 6 | 1620 | 3595770 | ||
BC037722.1 | - | 5 | 1431 | |||
BC039748.1 | - | 6 | 1631 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
