siRNA Details
Gene Symbol: 9330182L06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172706.3 | NP_766294.2 | 13, 14 | 1802 | ||
GenBank mRNA | AK034358.1 | - | 13, 14 | 1834 | ||
AK045969.1 | - | 13, 14 | 1836 | |||
AK048738.1 | - | 13, 14 | 1788 | |||
AK079644.1 | - | 12, 13 | 1733 | |||
AK133610.1 | - | 13, 14 | 1803 | |||
AK134808.1 | - | 13, 14 | 1864 | |||
AK161902.1 | - | 13, 14 | 1863 | |||
BC132290.1 | - | 13, 14 | 1778 | 40130567 | ||
BC138012.1 | - | 13, 14 | 1778 | 8861031 |
