siRNA Details
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF025653.1 | - | 6 | 566 | ||
AF034568.1 | - | 6 | 620 | |||
AK002922.1 | - | 6 | 669 | |||
AK079488.1 | - | 6 | 663 | |||
AK142016.1 | - | 6 | 663 | |||
AK150497.1 | - | 6 | 673 | |||
AK151463.1 | - | 6 | 687 | |||
AK160007.1 | - | 6 | 675 | |||
AK160706.1 | - | 6 | 653 | |||
AK169305.1 | - | 6 | 639 | |||
BC043657.1 | - | 6 | 638 | 5343539 |

More Information
Other Species Targeted

