siRNA Details
Gene Symbol: Ncoa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242558.1 | NP_001229487.1 | 3 | 388 | ||
NM_019825.3 | NP_062799.2 | 3 | 388 | |||
GenBank mRNA | AF216186.1 | - | 2 | 279 | ||
AK144338.1 | - | 5 | 627 | |||
AK171864.1 | - | 3 | 549 | |||
BC031113.1 | - | 3 | 392 | |||
BC083187.1 | - | 3 | 388 | 30544739 | ||
JF707634.1 | - | 2 | 247 | |||
JF707635.1 | - | 2 | 247 | |||
JF707636.1 | - | 2 | 247 |
