siRNA Details
Gene Symbol: Cks1brt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037922.3 | NP_001033011.1 | 2 | 1348 | ||
GenBank mRNA | AK144293.1 | - | 2 | 1349 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cks1brt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037922.3 | NP_001033011.1 | 2 | 1348 | ||
GenBank mRNA | AK144293.1 | - | 2 | 1349 |