siRNA Details
Gene Symbol: Sdf2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022324.3 | NP_071719.1 | 3 | 519 | ||
GenBank mRNA | AB043006.1 | - | 3 | 459 | ||
BC023290.1 | - | 3 | 473 | |||
BC053425.1 | - | 3 | 501 | 6508268 | ||
BC053438.1 | - | 3 | 501 |
