siRNA Details
Gene Symbol: 2310061I04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033630.1 | NP_001028802.1 | 3, 4 | 684 | ||
GenBank mRNA | AK038622.1 | - | 2, 3 | 1536 | ||
AK145087.1 | - | 3, 4 | 638 | |||
AK172070.1 | - | 3, 4 | 601 | |||
BC100355.1 | - | 2, 3 | 313 | 30527188 | ||
BC117035.1 | - | 2, 3 | 344 | |||
BC117039.1 | - | 2, 3 | 344 | |||
BC144744.1 | - | 2, 3 | 344 | |||
BQ444983.1 | - | 2, 3 | 127 |
