siRNA Details
Gene Symbol: Spata1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027617.3 | NP_081893.1 | 11 | 1245 | ||
GenBank mRNA | BC139279.1 | - | 10 | 1158 | 8862301 | |
BC139280.1 | - | 10 | 1158 | 8862302 | ||
BC171970.1 | - | 10 | 1156 |
