siRNA Details
Gene Symbol: Otud3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028453.1 | NP_082729.1 | 3 | 568 | ||
GenBank mRNA | AK014098.1 | - | 3 | 610 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Otud3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028453.1 | NP_082729.1 | 3 | 568 | ||
GenBank mRNA | AK014098.1 | - | 3 | 610 |