siRNA Details
Gene Symbol: VAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199131.2 | NP_954582.1 | 2, 3 | 679 | ||
GenBank mRNA | AK127095.1 | - | 2, 3 | 676 | ||
BC101694.1 | - | 2, 3 | 463 | 8069200 | ||
BC101696.1 | - | 2, 3 | 463 | 8069202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
