siRNA Details
Gene Symbol: Fst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012561.1 | NP_036693.1 | 4, 5 | 718 | ||
XM_006231954.2 | XP_006232016.1 | 4, 5 | 1078 | |||
GenBank mRNA | FQ220872.1 | - | 2, 3 | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
