siRNA Details
Gene Symbol: Grik4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012572.1 | NP_036704.1 | 13, 14 | 1665 | ||
GenBank mRNA | U08257.1 | - | 13, 14 | 1665 | ||
X59996.1 | - | 13, 14 | 1666 |

More Information
Other Species Targeted

Gene Ontology Categories:
