siRNA Details
Gene Symbol: Il6r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017020.3 | NP_058716.2 | 5 | 757 | ||
XM_006232587.2 | XP_006232649.1 | 5 | 2191 | |||
XM_006232588.2 | XP_006232650.1 | 5 | 2190 | |||
XM_008761127.1 | XP_008759349.1 | 5 | 2186 | |||
GenBank mRNA | M58587.1 | - | 5 | 845 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
