siRNA Details
Gene Symbol: Mme | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012608.2 | NP_036740.1 | 10 | 1023 | ||
XM_006232437.2 | XP_006232499.1 | 10 | 1059 | |||
GenBank mRNA | BC085753.1 | - | 10 | 1023 | 7110842 | |
M15944.1 | - | 9 | 973 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
