siRNA Details
Gene Symbol: Slc9a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173098.1 | NP_775121.1 | 1, 2 | 730 | ||
XM_006244738.2 | XP_006244800.1 | 1, 2 | 896 | |||
XM_008766988.1 | XP_008765210.1 | 1, 2 | 896 | |||
XM_008766989.1 | XP_008765211.1 | 2, 3 | 799 | |||
XM_008766990.1 | XP_008765212.1 | 2, 3 | 672 | |||
GenBank mRNA | M85301.1 | - | 1, 2 | 730 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
