siRNA Details
Gene Symbol: Sds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053962.3 | NP_446414.3 | 8 | 891 | ||
XM_008769213.1 | XP_008767435.1 | 7 | 1389 | |||
GenBank mRNA | BC088110.1 | - | 8 | 891 | 7323391 | |
J03863.1 | - | 8 | 1089 | |||
Y00752.1 | - | 8 | 908 |
