siRNA Details
Gene Symbol: Htr2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012765.3 | NP_036897.2 | 6 | 1584 | ||
XM_008773402.1 | XP_008771624.1 | 6 | 1950 | |||
GenBank mRNA | CB613191.1 | - | 1 | 97 | ||
U35315.1 | - | 6 | 1488 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
