siRNA Details
Gene Symbol: Gamt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012793.2 | NP_036925.2 | 3 | 430 | ||
GenBank mRNA | FM043834.1 | - | 3 | 403 | ||
FQ210612.1 | - | 3 | 353 | |||
FQ225896.1 | - | 3 | 430 | |||
FQ227114.1 | - | 3 | 430 | |||
J03588.1 | - | 3 | 428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
