siRNA Details
Gene Symbol: Acsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012820.1 | NP_036952.1 | 19 | 1914 | ||
XM_006253122.1 | XP_006253184.1 | 20 | 2574 | |||
XM_006253123.1 | XP_006253185.1 | 20 | 2115 | |||
XM_006253124.2 | XP_006253186.1 | 20 | 2034 | |||
XM_006253125.2 | XP_006253187.1 | 20 | 2194 | |||
XM_006253126.1 | XP_006253188.1 | 20 | 2040 | |||
XM_006253127.1 | XP_006253189.1 | 20 | 2005 | |||
XM_008771254.1 | XP_008769476.1 | 19 | 1940 | |||
GenBank mRNA | D90109.1 | - | 19 | 1914 | ||
FQ217275.1 | - | 19 | 1884 | |||
FQ219468.1 | - | 3 | 215 |
